Galliumantimonid, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ quality level = MQ100

SIAL651478-1EAEA 1820 EUR
SIAL651478-1EA
Galliumantimonid, Sigma-Aldrich®
Gallium antimonide
Formel: GaSb
Molecular Weight: 191,48 g/mol
Schmelzpunkt: 712 °C
Dichte: 5,5…5.7 g/cm³ (20 °C)
MDL: MFCD00016101
CAS-Nummer: 12064-03-8
EINECS: 235-058-8
UN: 1549
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR