4-Iodbiphenyl, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1-Jod-4-phenylbenzol
, 1-Iod-4-phenylbenzol
M-Clarity™ quality level = MQ100
SIAL637769-25GEA
175
EUR
SIAL637769-25G
SIAL637769-5G
4-Iodbiphenyl, Sigma-Aldrich®
4-Iodbiphenyl
Formel:
C₁₂H₉I Molecular Weight: 280,11 g/mol Siedepunkt: 320 °C (1013 hPa) Schmelzpunkt: 112 °C Dichte: 1,584 g/cm³ (20 °C) |
MDL:
MFCD00019028 CAS-Nummer: 1591-31-7 EINECS: 216-469-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...