α,α-Dimethoxytoluol

Lieferant: FLUOROCHEM LTD (CHEMPUR)
Achtung

Synonyme: Benzaldehyddimethylacetal

FLUO018193-25GEA 35.4 EUR
FLUO018193-25G
α,α-Dimethoxytoluol
α,α-Dimethoxytoluol
Formel: C₉H₁₂O₂
Molecular Weight: 152,19 g/mol
Siedepunkt: 198 °C (1013 hPa)
Dichte: 1,01 g/cm³ (20 °C)
MDL: MFCD00008491
CAS-Nummer: 1125-88-8
EINECS: 214-413-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR