ω-Brom-3-cyanacetophenon

Lieferant: Thermo Scientific

Synonyme: 3-(Bromacetyl)benzonitrilBrommethyl-m-cyanphenylketonα-Brom-3-cyanacetophenon3-Cyanphenacylbromid2-Brom-3'-cyanacetophenon

MAYBSPB07593DAEA 0 EUR
MAYBSPB07593DA MAYBSPB07593.0003
ω-Brom-3-cyanacetophenon
ω-Brom-3-cyanacetophenon
Formel: C₉H₆BrNO
Molecular Weight: 224,06 g/mol
MDL: MFCD00833267
CAS-Nummer: 50916-55-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR