5-Hydroxyisophthalsäure, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
5-Hydroxy-1,3-benzoldicarbonsäure
M-Clarity™ quality level = MQ100
SIAL311278-100GEA
57.8
EUR
SIAL311278-100G
5-Hydroxyisophthalsäure, Sigma-Aldrich®
5-Hydroxyisophthalsäure
Formel:
C₈H₆O₅ Molecular Weight: 182,13 g/mol Siedepunkt: 386 °C (1013 hPa) Schmelzpunkt: 298…301 °C |
MDL:
MFCD00002515 CAS-Nummer: 618-83-7 EINECS: 210-565-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...