4-Methoxyisophthalsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL528277-5GEA 218 EUR
SIAL528277-5G
4-Methoxyisophthalsäure, Sigma-Aldrich®
4-Methoxyisophthalsäure
Formel: C₉H₈O₅
Molecular Weight: 196,16 g/mol
MDL: MFCD06208422
CAS-Nummer: 2206-43-1
EINECS: 218-618-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR