5-Chlor-2-iodanilin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL685208-5GEA 54.6 EUR
SIAL685208-5G
5-Chlor-2-iodanilin, Sigma-Aldrich®
5-Chlor-2-iodanilin
Formel: C₆H₅ClIN
Molecular Weight: 253,47 g/mol
MDL: MFCD06738960
CAS-Nummer: 6828-35-9
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR