Ethyl-trans-cinnamat, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ quality level = MQ100

SIAL112372-100GEA 24.1 EUR
SIAL112372-100G
Ethyl-trans-cinnamat, Sigma-Aldrich®
Ethyl-trans-cinnamat
Formel: C₁₁H₁₂O₂
MDL: MFCD00009189
CAS-Nummer: 103-36-6
EINECS: 203-104-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR