Organic analytical standard, stearyl stearate, Supelco®
Lieferant: Merck
46408-100MGEA 107 EUR
46408-100MG
Organic analytical standard, stearyl stearate, Supelco®
Standards Organische Standards
Stearyl stearate may be used as a model lipid to stain contact lenses in evaluating the lipid-degrading ability of the cholesterol esterase enzyme produced from Pseudomonas aeruginosa.
Bitte beachten Shelf life: Limited shelf life, expiry date on the label.