Isobutylacetoacetat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-OxobuttersäureisobutylesterAcetessigsäureisobutylester

M-Clarity™ quality level = MQ100

SIAL00483-100MLEA 111 EUR
SIAL00483-100ML
Isobutylacetoacetat, Sigma-Aldrich®
Isobutylacetoacetat
Formel: C₈H₁₄O₃
Molecular Weight: 158,2 g/mol
MDL: MFCD00059349
CAS-Nummer: 7779-75-1
EINECS: 231-937-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR