Lutetiumtrinitrat Hydrat, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL542067-5GEA
333
EUR
SIAL542067-5G
Lutetiumtrinitrat Hydrat, Sigma-Aldrich®
Lutetiumtrinitrat Hydrat
Formel:
LuN₃O₉·H₂O Molecular Weight: 379 g/mol |
MDL:
MFCD00150421 CAS-Nummer: 100641-16-5 EINECS: 233-241-7 UN: 1477 ADR: 5.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...