L(+)-Lysin Monohydrochlorid

Synonyme: L(+)-Lysin HydrochloridH-Lys-OH.HCl(S)-(+)-2,6-Diaminocapronsäure Hydrochlorid(S)-(+)-Lysin Monohydrochlorid(S)-(+)-Lysin Hydrochlorid(S)-(+)-2,6-Diaminohexansäure Hydrochlorid(S)-(+)-2,6-Diaminocapronsäure Monohydrochlorid(S)-(+)-2,6-Diaminohexansäure Monohydrochlorid

EDQML0900000EA 235 EUR
EDQML0900000
L(+)-Lysin Monohydrochlorid
L(+)-Lysin Monohydrochlorid
Formel: H₂N(CH₂)₄CH(NH₂)CO₂H·HCl
Molecular Weight: 182,65 g/mol
Schmelzpunkt: 263…264 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00064564
CAS-Nummer: 657-27-2
EINECS: 211-519-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR