Concentric nebulizer/Scott spray chamber end cap sssembly for Avio and Optima
Lieferant: PerkinElmer
PERKN0680504EA 0 EUR
PERKN0680504
Concentric nebulizer/Scott spray chamber end cap sssembly for Avio and Optima
Zerstäuber
This end cap assembly, which connects concentric GemCone™ and similar nebulizers to Ryton Scott spray chambers, is used with Avio® 200/500 ICP-OES, and Optima® 2x00 DV/4x00 DV/5x00 DV/7x00 DV/8x00 ICP-OES series instruments.
Lieferumfang: This assembly includes: Nebuliser O-Ring (1), Knurled Screw (2), Insertion Ring (1), O-Ring, 34,29 mm Int.Ø, (1), Nut (1), End Cap (1), End Cap Washer (1).