MERC1.50691
1.50691.1000EA 110 EUR
1.50691.1000
MERC1.50691
3S is an intelligent packaging solution protected with an originality seal. The cap provides information via Radio-frequency identification or RFID, enabling automated data transfer between the reagent and titration instrument. Intelligent communication for improved performance. 3S Reagents are compatible with Metrohm’s OMNIS titration platform through 3S Adapter Technology.
  • RFID-based identification of reagent—automated and error-free
  • Complete traceability of the analyses provided
  • Aspiration tube already integrated into the bottle
  • Unique opening procedure minimizes risk of contamination