Autosampler aluminum sample pans
Lieferant: Perkin Elmer
PERKB0143017EA 0 EUR
PERKB0143017
Autosampler aluminum sample pans
Schalen/Schüsseln/Töpfe Instrumentenschalen
Autosampler sample pans are used to prepare sample materials for PerkinElmer DSC Autosamplers. These pans are sealed and may also be used for non-robotic systems.
  • Pans have an operating range down to –170 °C
  • Pans can withstand a pressure of 1 bar
  • Pan thickness is 0,1 mm
These aluminum sample pans can also be used with crimper (B0139005) and require cover (B0143003).