Conical bottom autosampler tubes with graduation
Lieferant: Perkin Elmer
PERKB0193233EA 0 EUR
PERKB0193233
Conical bottom autosampler tubes with graduation
Röhrchen Reagenzgläser- und Probenröhrchen
These standard translucent polypropylene tubes are designed to provide excellent chemical resistance ensuring an accurate analysis.
  • Compatible with PerkinElmer Autosamplers for AA, ICP-OES and ICP-MS instruments
  • Tubes are graduated for ease-of-use
  • Writing patch area for sample identification
These are the standard autosampler tubes for the PerkinElmer S10 Autosampler.