Germanium Standard for AAS, TraceCERT®, Supelco®
Lieferant: Merck
TraceCERT®
92685-250MLEA 29.4 EUR
92685-250ML
Germanium Standard for AAS, TraceCERT®, Supelco®
Standards Kationischer Standards Germanium-Standards
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34)