Organic reference standard, DL-3-Ureidoisobutyric acid, ≥99,0% (HPLC), Supelco®
Lieferant: Merck
74005-10MGEA 62.7 EUR
74005-10MG
Organic reference standard, DL-3-Ureidoisobutyric acid, ≥99,0% (HPLC), Supelco®
Standards Organische Standards
3-Ureidoisobutyrate is increased in the urine of patients with beta-ureidopropionase deficiency and can be used to predict enzyme deficiencies in pyrimidine metabolism of individual phenotypes, when associated with a risk for severe toxicity against the anti-neoplastic agent 5-fluorouracil.