Organic reference standards, (+)-Abscisic acid, analytical standard, Supelco®
Lieferant: Merck
90769-25MGEA 63.3 EUR
90769-25MG
Organic reference standards, (+)-Abscisic acid, analytical standard, Supelco®
Standards Organische Standards
Plant hormone and growth regulator that is involved in several physiological mechanisms including seed dormancy, leaf abscission, stomatal movement, and plant stress responses.
Through complex interactions with several intracellular signaling systems, it can regulate the expression of hundreds of plant genes.