Alkane Standard Mixture for performance tests of GC-systems, C10; C20 - C40 (all even), analytical standard, Supelco®
Lieferant: Merck
94234-10MLEA 411 EUR
94234-10ML
Alkane Standard Mixture for performance tests of GC-systems, C10; C20 - C40 (all even), analytical standard, Supelco®
Standards Mineralölindustrie-Standards