NHI-D FAME mix, analytical standard, Supelco®
Lieferant: Merck
O8381-1AMPEA 127 EUR
O8381-1AMP
NHI-D FAME mix, analytical standard, Supelco®
Standards Lebensmittel-Standards
Standards: Analytes: Methyl myristate 11.8 wt. %; Methyl oleate 44.6 wt. %; Methyl palmitate 23.6 wt. %; Methyl palmitoleate 6.9 wt. %; Methyl stearate 13.1 wt. %
Verpackung: Storage temperature: –20°C 