Pesticide reference standard, Imazalil, PESTANAL®, analytical standard, Supelco®
Lieferant: Merck
PESTANAL®
32007-100MGEA 63.4 EUR
32007-100MG
Pesticide reference standard, Imazalil, PESTANAL®, analytical standard, Supelco®
Standards Organische Standards
Utilised as a reference standard in direct analysis in real time mass spectrometry (DART-MS) technique for determination of xenobiotics from fruit samples.
  • Imazalil is a fungicide
  • Used to avoid decay during postharvest
  • Helps to control fungal growth in fruits, vegetables and ornamental plants
It may be used as reference standard in flow injection photoinduced chemiluminescence method for determination of Imazalil concentration from water samples.