Pesticide reference standard, Chlorantraniliprole, PESTANAL®, analytical standard, Supelco®
Lieferant: Merck
PESTANAL®
32510-25MGEA 103 EUR
32510-25MG
Pesticide reference standard, Chlorantraniliprole, PESTANAL®, analytical standard, Supelco®
Standards Organische Standards
Chlorantraniliprole is a novel anthranilic diamide insecticide belonging to the class of selective ryanodine receptor agonists. It is a selective and potent activator of insect ryanodine receptors (RyRs) that are critical for muscle contraction. It is very efficient for controlling lepidopteran insect pests, as well as some species in the orders Diptera, Hemiptera, and Coleoptera.
Chlorantraniliprole activates the RyRs in insects thereby affecting calcium homeostasis by unregulated release of internal calcium in the cell, leading to feeding cessation, muscle paralysis, lethargy, and ultimately death of the insect.