C1-C6 n-Paraffins, each component in nitrogen, analytical standards, Supelco®
Lieferant: Merck
501786.EA 0 EUR
501786. 23444. 501778. 501859. 330300. 501840.
C1-C6 n-Paraffins, each component in nitrogen, analytical standards, Supelco®
Standards Mineralölindustrie-Standards
Multi-component gas mixture suitable for GC calibration.
  • A certificate of analysis is included.
  • Packaged in lightweight, non-refillable, recyclable containers.
  • Portable, convenient, and easy to use.
  • Manufactured gravimetrically for maximum accuracy.
  • NIST traceable.