LEUCOGNOST™ Leukemia Detection Kit, Sigma-Aldrich®
Lieferant: Merck
1.16302.0002EA 189 EUR
MERC1.16302.0002
LEUCOGNOST™ Leukemia Detection Kit, Sigma-Aldrich®
LEUCOGNOST™ leukemia detection kit provide the user with all the necessary reagents and easy-to-follow staining
instructions.
LEUCOGNOST™ leukemia detection kit now enable differential analysis for leukaemia to be carried out in
international FAB classes by smaller laboratories, hence making the technique no longer restricted to larger diagnostic
centers.
The staining kit is sufficient for 12 stainings with up to 16 slides.
A fixing reagent for diagnosis with LEUCOGNOST™ leukemia detection kit is available: see LEUCOGNOST™ Fixing Mixture - code 112327 (500 ml)