Hygromycin B-Lösung 50 mg/ml in HEPES (aus Streptomyces species) ≥90% (durch HPLC) auf Zellkulturen getestet, Calbiochem®, Millipore®
|
Gefahr
|
400053-20EA
284
EUR
400053-20
400053-5
Hygromycin B-Lösung 50 mg/ml in HEPES (aus Streptomyces species) ≥90% (durch HPLC) auf Zellkulturen getestet, Calbiochem®, Millipore®
Hygromycin B
Hygromycin B, is isolated from Streptomyces sp., sterile-filtered solution in 25 mM HEPES and cell culture-tested.
Hygromycin B is an aminoglycoside antibiotic that inhibits the growth of prokaryotic and eukaryotic microorganisms and mammalian cells. Inhibits protein synthesis by interfering with the translocation of the 70S ribosome and causes misreading of mRNA.
Formel:
C₂₀H₃₇N₃O₁₃ Molecular Weight: 527,5 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06795479 CAS-Nummer: 31282-04-9 UN: 3172 ADR: 6.1,III |
Spezifikation Testergebnisse
Purity | ≥90% (HPLC) |