Pure water viscosity standard, UKAS ISO 17025 and ISO Guide 34 certified, Supelco®
Lieferant: Merck
SIALVISCWAT-100MLEA 212 EUR
SIALVISCWAT-100ML
Pure water viscosity standard, UKAS ISO 17025 and ISO Guide 34 certified, Supelco®
Standards Viskosimetrie-Standards
Standard is certified in strict accordance with ASTM D2162, the primary method for viscosity standards manufacture, for temperatures at 20 °C, 25 °C and 37 °C.
Paragon ISO 17025 / ISO Guide 34 certified pure water viscosity standards are specifically designed, tested and certified for the calibration, verification and certification of glass capillary viscometers and other viscosity measuring equipment where the precise control of temperature is required.
Standards: This product is a certified reference material (CRM) certified to International Standards BS EN ISO / IEC 17025 and ISO Guide 34 under UKAS accreditation.