Organic reference standard, neat, Dimethyl sulfone, (CRM), standard for quantitative NMR, TraceCERT®, Supelco®
Lieferant: Merck
TraceCERT®
41867-1GEA 111 EUR
41867-1G
Organic reference standard, neat, Dimethyl sulfone, (CRM), standard for quantitative NMR, TraceCERT®, Supelco®
Standards Organische Standards
Dimethyl sulfone is a solvent, which can dissolve a wide range of solutes and can be generally used for high temperature reactions.
Dimethyl sulfone has been used as internal standard, for the nuclear magnetic resonance spectrum studies of cocrystals containing API carbamazepine, synthesized using mechanochemical method of synthesis.
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34). This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.