Neutralrot 1% in wässriger Lösung, Clin-Tech
Lieferant: KLINIPATH (PL)
Synonyme:
Basisrot 5
, 2-Amino-8-(dimethylamino)-3-methylphenaziniumchlorid
, N8,N8,3-Trimethyl-2,8-phenazindiamin Monohydrochlorid
VWRK620225EA
21.4
EUR
VWRK620225
Neutralrot 1% in wässriger Lösung, Clin-Tech
Neutralrot
Formel:
C₁₅H₁₇ClN₄ Molecular Weight: 288,78 g/mol |
MDL:
MFCD00012651 CAS-Nummer: 553-24-2 |
![]() |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...