Flame thermionic detector (FTD)
Lieferant: Shimadzu
SHIM221-73332-48EA 0 EUR
SHIM221-73332-48
Flame thermionic detector (FTD)
Chromatographie-Detektoren
This detector features a digital pressure controller and is applicable for GC-2010/2010 Plus.
Lieferumfang: Includes one piece of collector as a standard accessory.
Bitte beachten FTD requires hydrogen and air as supply gas. Supply air should be from a high-purity air cylinder. It is not recommended to use air from a compressor.