Connector, VarioPin
Lieferant: Mettler-Toledo
VarioPin
METN52200241EA 0 EUR
METN52200241
Connector, VarioPin
Steckdosen
This connector fits with shielded coaxial cables (standard and high temperature).