Carbon black, mesoporös, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
SIAL702110-5GEA
272
EUR
SIAL702110-5G
Carbon black, mesoporös, Sigma-Aldrich®
Carbon black
Carbon with hydrophilic pore surface.
- Particle size: 5 to 50 μm (SEM)
- Pore size: >0.2 cm³/g mesoporosity
- Specific surface area: >50 m²/g (BET)
Starbons: Cooking up nanostructured mesoporous materials.
Formel:
C Molecular Weight: 12,01 g/mol Siedepunkt: 4827 °C Schmelzpunkt: 3654… 3697 °C |
MDL:
MFCD00133992 CAS-Nummer: 1333-86-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...