Dextran, M.W. 60000 - 90000, ultrarein

J14495.22EA 80.7 EUR
J14495.22 J14495.A1
Dextran, M.W. 60000 - 90000, ultrarein
Dextran
Formel: (C₆H₁₀O₅)n
Lagertemperatur: Raumtemperatur
MDL: MFCD00130935
CAS-Nummer: 9004-54-0
EINECS: 232-677-5

Bestellen

Spezifikation Testergebnisse

Residue on Ignition ≤1.9%
Loss on Drying ≤7.0%
pH (6% w/v in KCl) 4.5 - 7.0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR