Chemical dispensing station, Chemstat 2.0
Lieferant: Sermo Industrialisation
720-2942EA 96210 EUR
720-2942
Chemical dispensing station, Chemstat 2.0
Dunstabzugshauben und Verkleidungen Chemikalien-Abzüge
Thanks to the new Chemstat 2,0, the search for safe and correct distribute and dilute of solvents is over.
  • Extraction at the bottom for formaldehyde and at the top for solvents
  • Archiving each action and user for traceability
  • Ergonomic extendable carriers in the cabinet up to 60 liters
  • Electric safety valve on the waste container
  • Height-adjustable distribution faucets
Standards: Anti-fire and explosive cabinet according to EN 14470-1 and TRGS 510.
Bestellinformation: Available with two or three dispensers that can all dilute.