3-Amino-2,5-dichlorpyridin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL706817-1GEA 65.3 EUR
SIAL706817-1G
3-Amino-2,5-dichlorpyridin, Sigma-Aldrich®
3-Amino-2,5-dichlorpyridin
Formel: C₅H₄Cl₂N₂
Molecular Weight: 163,01 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00204157
CAS-Nummer: 78607-32-6
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR