1,8-Dimethylnaphthalin, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL282537-1GEA
154
EUR
SIAL282537-1G
1,8-Dimethylnaphthalin, Sigma-Aldrich®
1,8-Dimethylnaphthalene
Formel:
C₁₂H₁₂ Molecular Weight: 156,23 g/mol Siedepunkt: 270 °C (1013 hPa) Schmelzpunkt: 60…62 °C Dichte: 1,14 g/cm³ (20 °C) |
CAS-Nummer:
569-41-5 EINECS: 209-314-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...