Digadoliniumtrioxid, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
Gadolinium(III)oxid
SIAL203297-5GEA
184
EUR
SIAL203297-5G
Digadoliniumtrioxid, Sigma-Aldrich®
Digadoliniumtrioxid
Formel:
Gd₂O₃ Molecular Weight: 362,5 g/mol Schmelzpunkt: 2410 °C Dichte: 7,407 g/cm³ (20 °C) |
MDL:
MFCD00011026 CAS-Nummer: 12064-62-9 EINECS: 235-060-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...