Ytterbium(III)fluorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ytterbiumtrifluorid

M-Clarity™ quality level = MQ100

SIAL432121-10GEA 188 EUR
SIAL432121-10G
Ytterbium(III)fluorid, Sigma-Aldrich®
Ytterbium(III)fluorid
Formel: F₃Yb
Molecular Weight: 230,04 g/mol
Siedepunkt: 2227 °C (1013 hPa)
Schmelzpunkt: 1163,85…1179.85 °C
Dichte: 8,2 g/cm³ (20 °C)
MDL: MFCD00049615
CAS-Nummer: 13760-80-0
EINECS: 237-354-2
UN: 3288
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR