Trioctylsilan

Lieferant: SILANE (PART OF CHEMPUR)
Achtung

SILNSILS20500.25EA 227 EUR
SILNSILS20500.25 SILNSILS20500.10 FLUOS20500.25G SILNSILS20500.100
Trioctylsilan
Trioctylsilan
Formel: C₂₄H₅₂Si
Molecular Weight: 368,76 g/mol
MDL: MFCD00053812
CAS-Nummer: 18765-09-8
EINECS: 242-559-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR