Propyltriethoxysilan, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL539317-25MLEA 0 EUR
SIAL539317-25ML
Propyltriethoxysilan, Sigma-Aldrich®
Propyltriethoxysilan
Formel: C₉H₂₂O₃Si
Molecular Weight: 206,36 g/mol
MDL: MFCD00026756
CAS-Nummer: 2550-02-9
EINECS: 219-842-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR