Natriumtetrafluoroborat
Lieferant: Honeywell Chemicals
SIAL01510-25KG-HEA
0
EUR
SIAL01510-25KG-H
HONE01510.9025
Natriumtetrafluoroborat
Sodium tetrafluoroborate
Formel:
BF₄Na Molecular Weight: 109,79 g/mol Schmelzpunkt: 384 °C Dichte: ∼2,47 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003515 CAS-Nummer: 13755-29-8 EINECS: 237-340-6 Merck Index: 12,08761 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...