2,4,6,8-Tetramethylcyclotetrasiloxan
Lieferant: SILANE (PART OF CHEMPUR)
Synonyme:
1,3,5,7-Tetramethylcyclotetrasiloxan
SILNSILS15975.100EA
207
EUR
SILNSILS15975.100
SILNSILS15975.5
SILNSILS15975.25
2,4,6,8-Tetramethylcyclotetrasiloxan
2,4,6,8-Tetramethylcyclotetrasiloxan
Formel:
C₄H₁₆O₄Si₄ Molecular Weight: 240,51 g/mol Siedepunkt: 142 °C (1013 hPa) Schmelzpunkt: –69 °C Dichte: 0,99 g/cm³ (20 °C) |
MDL:
MFCD00039567 CAS-Nummer: 2370-88-9 EINECS: 219-137-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...