Bromcyclododecan

Lieferant: Thermo Scientific

MAYBBTB09774.10EA 0 EUR
MAYBBTB09774.10 MAYBBTB09774.1
Bromcyclododecan
Bromcyclododecan
Formel: C₁₂H₂₃Br
Molecular Weight: 247,22 g/mol
CAS-Nummer: 7795-35-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR