2,3,5,6-Tetrachlorbiphenyl (PCB Nr. 65)
EHERE20006500EA
106
EUR
EHERE20006500
2,3,5,6-Tetrachlorbiphenyl (PCB Nr. 65)
2,3,5,6-Tetrachlorbiphenyl (PCB Nr. 65)
Formel:
C₁₂H₆Cl₄ Molecular Weight: 291,99 g/mol |
CAS-Nummer:
33284-54-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...