2,4,4',6-Tetrachlorbiphenyl (PCB Nr. 75)

Lieferant: EHRENSTORFER GMBH

C20007500
EHERC20007500EA 63 EUR
EHERC20007500
2,4,4',6-Tetrachlorbiphenyl (PCB Nr. 75)
2,4,4',6-Tetrachlorbiphenyl (PCB Nr. 75)
Formel: C₁₂H₆Cl₄
Molecular Weight: 291,99 g/mol
CAS-Nummer: 32598-12-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR