2,3,3',4',5-Pentachlorbiphenyl (PCB Nr. 107)
Lieferant: LGC Standards PROMOCHEM
PROOCIL-PCB-107-CEA
652
EUR
PROOCIL-PCB-107-C
2,3,3',4',5-Pentachlorbiphenyl (PCB Nr. 107)
2,3,3',4',5-Pentachlorbiphenyl (PCB Nr. 107)
Formel:
C₁₂H₅Cl₅ Molecular Weight: 326,44 g/mol |
CAS-Nummer:
70424-68-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...