2,2',3,4',5-Pentachlorbiphenyl (PCB Nr. 90)

EHERE20009000EA 0 EUR
EHERE20009000
2,2',3,4',5-Pentachlorbiphenyl (PCB Nr. 90)
2,2',3,4',5-Pentachlorbiphenyl (PCB Nr. 90)
Formel: C₁₂H₅Cl₅
Molecular Weight: 326,44 g/mol
CAS-Nummer: 68194-07-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR