2,6-Xylylisothiocyanat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,6-Dimethylphenylisothiocyanat

M-Clarity™ quality level = MQ100

SIAL476439-1GEA 24.6 EUR
SIAL476439-1G
2,6-Xylylisothiocyanat, Sigma-Aldrich®
2,6-Xylylisothiocyanat
Formel: C₉H₉NS
Molecular Weight: 163,24 g/mol
Siedepunkt: 67 °C (0,4 mmHg)
Dichte: 1,085 g/cm³ (20 °C)
MDL: MFCD00038715
CAS-Nummer: 19241-16-8
EINECS: 242-906-0
UN: 3334
ADR: 9,

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR