3,5-Dimethoxytoluol

Lieferant: Molekula
Achtung

Synonyme: 1,3-Dimethoxy-5-methylbenzol

MOLEM24183551EA 0 EUR
MOLEM24183551
3,5-Dimethoxytoluol
3,5-Dimethoxytoluol
Formel: C₉H₁₂O₂
Molecular Weight: 152,19 g/mol
MDL: MFCD00015435
CAS-Nummer: 4179-19-5
EINECS: 224-048-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR