3',4'-Difluoracetophenon, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1-(3,4-Difluorphenyl)ethan-1-on
M-Clarity™ quality level = MQ100
SIAL264288-5GEA
66.9
EUR
SIAL264288-5G
3',4'-Difluoracetophenon, Sigma-Aldrich®
3',4'-Difluoracetophenon
Formel:
C₈H₆F₂O Molecular Weight: 156,13 g/mol Siedepunkt: 207 °C (1013 hPa) Schmelzpunkt: 20 °C Dichte: 1,24 g/cm³ (20 °C) |
MDL:
MFCD00009891 CAS-Nummer: 369-33-5 EINECS: 206-717-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...